Learn More
Brady™ ISO Safety Awareness Signs
Product Code. 15384846
Change view
Click to view available options
Material:
Polyester
Polypropylene
Displayed Text:
CAUTION Hot Surface Pictogram
Doctor
Emergency exit (right)
Emergency phone
Eyewash Station
First Aid
Safety Shower
Unit Size:
Each
Pack of 7
13 product options available for selection
Product selection table with 13 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material Displayed Text unitSize
15384846 Polypropylene Doctor Each
13254359 Polyester CAUTION Hot Surface Pictogram Each
15634737 Polyester CAUTION Hot Surface Pictogram Pack of 7
15103299 Polyester Emergency exit (right) Each
15314856 Polyester Emergency phone Each
15374846 Polyester Doctor Each
15354846 Polyester First Aid Each
15364846 Polypropylene First Aid Each
15394846 Polyester Eyewash Station Each
15304856 Polypropylene Eyewash Station Each
15334856 Polyester Safety Shower Each
15344856 Polypropylene Safety Shower Each
15324856 Polypropylene Emergency phone Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 13 options available.
13 options
Product Code. 15384846 Supplier Brady™ Supplier No. 815242
Only null left
Add to basket
Add to basket
Help ensure safety at work and maximize productivity with these safety signs. Brady™ ISO Safety Awareness Signs are available in a variety of sizes. Standardized images reduce the risk of misunderstandings.
  • ISO 7010 Compliant
  • Indoor/outdoor sign
Specifications
Certifications/Compliance ISO 7010
Alternate Part Number PIC E009-250x250-PP-CRD/1
Material Polypropylene
Quantity 1 Ea.
Adhesive Type Non-adhesive
For Use With (Application) Pipe markers and safety signs
Height (Metric) 250 mm
Thickness 1.501 mm
Width (Metric) 250 mm
Temperature Range (Metric) -40°C to 120°C
Shape Square
Languages Global
Displayed Text Doctor
Product Type Safety Sign
Color White on Green