Learn More
Brady™ Laminated Polyester Radioactive Warning Sign
Specifications
Specifications
| Alternate Part Number | PIC 304-TRI 200-B7541 (BRANDED) |
| Material | Polyester |
| Quantity | 1 Ea. |
| Adhesive Type | Permanent Acrylic |
| Description | Laminated Polyester |
| For Use With (Application) | Used for indoor and outdoor safety signs |
| Height (Metric) | 200 mm |
| Max. Temperature (Metric) | 110°C |
| Width (Metric) | 200 mm |
| Shape | Triangular |
| Show More |