Learn More
Brady™ Laminated Polyester Radiation Laser Sign
Product Code. 11760194
Change view
Click to view available options
Displayed Text:
Laser beam warning
Warning of radioactive materials or radiation ioniserenden
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Displayed Text unitSize
11760194 Warning of radioactive materials or radiation ioniserenden Each
11780184 Laser beam warning Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11760194 Supplier Brady™ Supplier No. 223479
Only null left
Specifications
Alternate Part Number PIC 304-TRI 315-B7541
Material Polyester
Quantity 1 Ea.
Adhesive Type Permanent Acrylic
Description Laminated Polyester
For Use With (Application) Pipe markers and safety signs
Height (Metric) 315 mm
Max. Temperature (Metric) 110°C
Thickness 0.024 mm
Width (Metric) 315 mm
Shape Triangular
Displayed Text Warning of radioactive materials or radiation ioniserenden
Product Type Radiation Laser Sign
Color Black on Yellow