Learn More
Brady™ Laminated Polyester Prohibition Signs: Eating and Drinking Prohibited
Click to view available options
Diameter (Metric):
100 mm
200 mm
Unit Size:
Each
Specifications
Specifications
| Alternate Part Number | PIC 224-DIA 200-B7541 (BRANDED) |
| Diameter (Metric) | 200 mm |
| Shape | Round |