Learn More
Brady™ Laminated Polyester Mounted Anti-skid Tape
Product Code. 11721665
Click to view available options
Color:
Black
Red
Dimensions (L x W):
18 m x 100 mm
18 m x 150 mm
18 m x 25 mm
18 m x 300 mm
18 m x 50 mm
Unit Size:
1 roll
Product Code. 11721665
Brand: Brady™ 078199
Laminated Polyester Mounted Anti-Skid (anti-slip) Tape helps to prevent slipping and falling on wet or slippery floors.
  • The rugged polyester tape is adhesive backed and coated with thousands of particles of durable aluminium oxide grit.
  • Anti-Skid tape bonds permanently to clean, dry floors, withstands grease and oil and will work indoors or outdoors.
  • Anti-Skid tape is available in a variety of colors and sizes to suit all of your floor marking needs.
Specifications
Alternate Part Number ANTISKID TAPE RED 100MMX18M
Description Roll-Mounted Anti-Skid Tape - Red
Dimensions (L x W) 18 m x 100 mm
For Use With (Application) Traction on steps, slick floors, and egress pathway marking
Length (English) 59.1 ft.
Length (Metric) 18 m
Packaging Quantity 1 Roll
Temperature Range (English) -40°F to +158°F
Temperature Range (Metric) -40°C to +70°C
Thickness 0.162 mm
Width (English) 3.94 in.
Width (Metric) 100 mm
Adhesive Type Permanent Acrylic
Color Red
Material Polyester
Type Anti-skid Tape