Learn More
Brady™ Laminated Polyester Mandatory Signs
Product Code. 11700174
Change view
Click to view available options
Diameter (Metric):
100 mm
200 mm
310 mm
Displayed Text:
Use Breathing Protection
Use Face Shield
Use Full Protective
Use Overshoes
Use Protective Clothing
Use Protective Hood
Use eye protection
Use head protection safety
Wear Protective Footwear
Unit Size:
Each
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Displayed Text unitSize
11700174 310 mm Wear Protective Footwear Each
10161974 200 mm Use Breathing Protection Each
10613195 100 mm Use eye protection Each
10294844 100 mm Use Face Shield Each
10204854 200 mm Use Face Shield Each
10112264 200 mm Use Full Protective Each
10274794 200 mm Use head protection safety Each
10643385 100 mm Use Overshoes Each
10396144 200 mm Use Overshoes Each
10205094 200 mm Use Protective Clothing Each
10204564 100 mm Use Protective Hood Each
10451264 200 mm Use Protective Hood Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
Product Code. 11700174 Supplier Brady™ Supplier No. 222565
Only null left
Add to basket
Add to basket
Mandatory signs to assist with personal safety
  • Complies to NF X 08-003-1 and 08-003-3 N X and EU Directive 92/58/EEC
  • Resistant to chemicals; water; UV light; scratch/abrasion and temperatures up to 110°C
  • Specifications
    Alternate Part Number PIC 280-DIA 315-B7541
    Material Polyester
    Quantity 1 Ea.
    Adhesive Type Permanent Acrylic
    Description Laminated Polyester
    Diameter (Metric) 310 mm
    Max. Temperature (Metric) 110°C
    Thickness 0.024 mm
    Shape Round
    Displayed Text Wear Protective Footwear
    Product Type Mandatory Sign
    Color White on Blue