Learn More
Brady™ Laminated Polyester: ISO Safety Sign - Wear protective clothing
Product Code. 15604497
Change view
Click to view available options
Diameter (Metric):
50 mm
100 mm
200 mm
315 mm
Unit Size:
Each
Pack of 2
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) unitSize
15604497 50 mm Pack of 2
15614497 100 mm Each
15624497 200 mm Each
15634497 315 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 15604497 Supplier Brady™ Supplier No. 819312
Only null left
Add to basket
Add to basket
Sign compliant to ISO 7010.
  • Laminated polyester
  • For indoor and outdoor use
Specifications
Certifications/Compliance ISO 7010
Alternate Part Number PIC M010-Dia 050-PE-SHEET/2
Finish Gloss
Material Polyester
Quantity 2/Pk.
Adhesive Type Permanent Acrylic
Diameter (Metric) 50 mm
For Use With (Application) Indoor and Outdoor
Thickness 0.099 mm
Temperature Range (Metric) -40°C to 110°C
Shape Round
Languages Global
Color White on Blue