Learn More
Brady™ Laminated Polyester: ISO Safety Sign - Warning; Corrosive substance
Product Code. 15518850 Shop All Brady Products
Change view
Click to view available options
Width (Metric):
50 mm
100 mm
Height (Metric):
43 mm
87 mm
Unit Size:
Each
Pack of 7
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Width (Metric) Height (Metric) Unit Size
15518850 100 mm 87 mm Each
15313927 50 mm 43 mm Pack of 7
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15518850 Supplier Brady™ Supplier No. 829723
  • ONLINE EXCLUSIVE PRICE
    9.40€
    9.19€ / Pack of 1
    Save 0.21€
    2% Off
In Stock
Sign compliant to ISO 7010.
  • Laminated polyester
  • For indoor and outdoor use
Specifications
Certifications/Compliance ISO 7010
Alternate Part Number PIC W023-Tri 100-PE-SHEET/3
Finish Gloss
Material Polyester
Quantity 3/Pk.
Adhesive Type Permanent Acrylic
For Use With (Application) Indoor and Outdoor
Height (Metric) 87 mm
Thickness 0.099 mm
Width (Metric) 100 mm
Temperature Range (Metric) -40°C to 110°C
Shape Triangular
Languages Global
Color Black on Yellow