Learn More
Brady™ Laminated Polyester: ISO Safety Sign - Use this walkway
Product Code. 15664557
Click to view available options
Diameter (Metric):
50 mm
100 mm
200 mm
315 mm
Unit Size:
Each
Pack of 2
Product Code. 15664557
Brand: Brady™ 821401
Only null left
Add to basket
Add to basket
Sign compliant to ISO 7010.
  • Laminated polyester
  • For indoor and outdoor use
Specifications
Certifications/Compliance ISO 7010
Alternate Part Number PIC M024-Dia 200-PE-CRD/1
Finish Gloss
Material Polyester
Quantity 1 Ea.
Adhesive Type Permanent Acrylic
Diameter (Metric) 200 mm
For Use With (Application) Indoor and Outdoor
Thickness 0.099 mm
Temperature Range (Metric) -40°C to 110°C
Shape Round
Languages Global
Color White on Blue