Description
The Corning™ Lambda™ EliteMax Semi-automated Benchtop Pipettor can perform a variety of liquid handling tasks including:
- Plate filling from reagent reservoirs
- Serial dilutions, plate-to-plate transfers, tube-to-tube transfers, and plate replication
- Well-to-well transfers (cherry picking protocol for single-channel head only). Unit comes with a single-channel and 12-channel head installed.
- Accurately and precisely transferring liquids and compounds between microplates
- Transferring by row or by column
More complex applications can be performed such as sample preparation, timed reaction, PCR preparation, and cell-based assays set up.
Specifications
Specifications
| Accuracy | 20 μL < ±2.0%; 100 μL < ±1.5%; 200 μL < ±1.0% |
| Product Type | Semi-Automated Benchtop Pipettor |
| Device Holds | Single-channel, 12-channel, 24-channel and custom-channel heads |
| Channels | 1, 12 |
| Volume (Metric) | 1 to 200 μL |
| For Use With (Equipment) | Corning™ Lambda™ EliteMax pipet tips |
| Includes | Corning™ Lambda™ EliteMax Semi-automated Benchtop Pipettor, Touch screen controller with software and license, External power supply, Portrait adapter stainless steel plate, Power cords (US, EU, UK, CH), Instruction manual, Calibration certificate |
| Plate Type | 96-well standard, shallow, deep well, microcentrifuge, V-, U-, and flat-bottom |
| Precision | 20 μL <1.5% CV; 100 μL <1.0% CV; 200 μL <0.7% CV |
| Temperature (Metric) | 5°C to 40°C (relative humidity 80°C% up to 30°C or 45% relative humidity at 40°C) |
| Show More |
Product Title
Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.
Haben Sie Verbesserungsvorschläge?