Learn More
Novus Biologicals™ Lactoferrin Native Protein
Product Code. 18096768 Shop All Bio Techne Products
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
Product Code. 18096768
Brand: Novus Biologicals™ NBP212557100UG
Highly purified. Generating reliable and reproducible results.
Native Lactoferrin The Lactoferrin Native Protein is derived from Human neutrophil. The Lactoferrin Native Protein has been validated for the following applications: Western Blot, SDS-Page.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) SDS-PAGE, Western Blot
Formulation 100 mM Tris-HCl (pH 8.0), 300 mM NaCl
Gene ID (Entrez) 4057
Molecular Weight (g/mol) 78 kDa
Quantity 100 μg
Source Human neutrophil
Storage Requirements Store at -20 to -80C. Avoid freeze-thaw cycles.
Gene Alias EC 3.4.21, EC 3.4.21.-, GIG12, growth-inhibiting protein 12, HLF2, Lactoferrin, lactotransferrin, LF, neutrophil lactoferrin, talalactoferrin
Gene Symbol LTF
Buffer 100 mM Tris-HCl (pH 8.0), 300 mM NaCl
Purity or Quality Grade >95%, by SDS-PAGE