Learn More
Lacto-N-neotetraose, TRC
Product Code. 30004887
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30004887 10 mg 10mg
30137771 5 mg 5mg
30134034 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30004887 Supplier Toronto Research Chemicals Supplier No. TRCL30011810MG
 Restricted Item
Estimated Shipment: 14-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 13007-32-4
Molecular Formula C26H45NO21
Molecular Weight (g/mol) 707.63
Synonym O-beta-D-galactopyranosyl-(1-4)-O-2-(acetylamino)-2-deoxy-beta-D-glucopyranosyl-(1-3)-O-beta-D-galactopyranosyl-(1-4)-D-Glucose, O-beta-D-galactopyranosyl-(1-4)-O-2-acetamido-2-deoxy-beta-D-glucopyranosyl-(1-3)-O-beta-D-galactopyranosyl-(1-4)-D-Glucopyranose (8CI)Lacto-N-neotetraose (6CI), O-beta-D-Galactopyranosyl-(1-4)-O-2-(acetylamino)-2-deoxy-beta-D-glucopyranosyl-(1-3)-O-beta-D-galactopyranosyl-(1-4)-D-glucose, LNnt, N-Neotetraose
IUPAC Name N-[(2S,3R,4R,5S,6R)-2-[(2R,3S,4S,5R,6S)-3,5-dihydroxy-2-(hydroxymethyl)-6-[(2R,3S,4R,5R)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxyoxan-4-yl]oxy-4-hydroxy-6-(hydroxymethyl)-5-[(2S,3R,4S,5R,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxyoxan-3-yl]acetamide
SMILES CC(=O)N[C@@H]1[C@@H](O)[C@H](O[C@@H]2O[C@H](CO)[C@H](O)[C@H](O)[C@H]2O)[C@@H](CO)O[C@H]1O[C@H]3[C@@H](O)[C@@H](CO)O[C@@H](O[C@H]4[C@H](O)[C@@H](O)C(O)O[C@@H]4CO)[C@@H]3O
Specifications
Quantity 10 mg
InChI Formula InChI=1S/C26H45NO21/c1-6(32)27-11-14(35)20(46-25-18(39)15(36)12(33)7(2-28)43-25)10(5-31)45-24(11)48-22-13(34)8(3-29)44-26(19(22)40)47-21-9(4-30)42-23(41)17(38)16(21)37/h7-26,28-31,33-41H,2-5H2,1H3,(H,27,32)/t7-,8-,9-,10-,11-,12+,13+,14-,15+,16-,17-,18-,19-,20-,21-,22+,23?,24+,25+,26+/m1/s1
Formula Weight 707.2484
Chemical Name or Material Lacto-N-neotetraose