missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LACTB2 Monoclonal Antibody (OTI4E7), TrueMAB™, OriGene
Mouse Monoclonal Antibody
242.00€ - 540.87€
Specifications
Antigen | LACTB2 |
---|---|
Clone | OTI4E7 |
Concentration | 0.43 mg/mL |
Applications | Flow Cytometry, Western Blot |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17507271
|
Origene Technologies
TA502583S |
30 μL |
242.00€
30µL |
Estimated Shipment: 27-05-2024
Log in to see stock. |
|||||
17587261
|
Origene Technologies
TA502583 |
100 μL |
540.87€
100µL |
Estimated Shipment: 27-05-2024
Log in to see stock. |
|||||
Description
LACTB2 Monoclonal antibody specifically detects LACTB2 in Human samples. It is validated for Flow Cytometry, Western BlotSpecifications
LACTB2 | |
0.43 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
Q53H82 | |
IgG1 | |
-20° C, Avoid Freeze/Thaw Cycles |
OTI4E7 | |
Flow Cytometry, Western Blot | |
Unconjugated | |
Mouse | |
Human | |
51110 | |
Primary |