missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Lac-Phe

Product Code. 30281232
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 30281232

Brand: Tocris Bioscience™ 7942/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Metabolite generated by cytosol nonspecific dipeptidase (CNDP2) during intense exercise

Lac-Phe is a metabolite generated from lactate and phenylalanine by cytosolic non-specific dipeptidase (CNDP2) during intense exercise. Lac-Phe reduces food intake in diet-induced obese mice without affecting movement or energy expenditure. When administrated chronically, Lac-Phe decreases adiposity and body weight and improves glucose homeostasis.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-[(2S)-2-Hydroxy-1-oxopropyl]-L-phenylalanine
CAS 183241-73-8
Quantity 50 mg
Target DNA, RNA and Protein Synthesis Compounds
Molecular Formula C12H15NO4
Purity 0.95
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.