Learn More
Grant Instruments™ Labwise™ Software
Product Code. 11476457
Change view
Click to view available options
For Use With (Equipment):
Grant Optima TX150, TXF200, LT ecocool Baths and Circulators
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
11476457 Grant Optima TX150, TXF200, LT ecocool Baths and Circulators Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11476457 Supplier Grant Instruments™ Supplier No. LABWISE
Add to basket
Add to basket
Labwise™ is a powerful and convenient software package for programming, controlling and recording key parameters of high performance baths and circulators in the Grant Optima range via a PC.
  • Full control of set-up, multi-segment programming and data logging for heating and cooling
  • Real-time status windows with graphic display including zooming and scaling
  • Operates in combination with Grant Optima™ TX150, TXF200 and LT ecocoolTM series baths and circulators
  • Enables easy control of relays and remote switching devices, including multiple segments
TRUSTED_SUSTAINABILITY
Specifications
Type Software
For Use With (Equipment) Grant Optima TX150, TXF200, LT ecocool Baths and Circulators