Learn More
Kirsch Philipp™ Laboratory Freezer
Product Code. 10196810
Change view
Click to view available options
Capacity (Metric) Refrigerator:
500 L
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Refrigerator Unit Size
10196810 500 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10196810 Supplier Kirsch Philipp™ Supplier No. 10050
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Re-circulating air cooling, a rapid defrosting system, quick-freeze function.
  • Re-circulating air cooling
  • Optimized ventilation plates to minimize interior temperature differences
  • Rapid freezing cycle
  • LED temperature display
  • Automatical defrosting and melt water evaporation
  • Easy accessible control panel with key switch
  • Micro-processor controlled temperature and thawing regulation to maximize temperature stability
  • Minimum/maximum temperature memory: optical and acoustical alarm
  • RS-485 interface for computer based temperature documentation
  • Potential free contact for alarm transmission
  • Lockable door
Specifications
Type Laboratory Freezer
Capacity (Metric) Refrigerator 500 L
Temperature Range (Freezer) -5°C to -20°C
Shelves 5 Adjustable
Capacity (Metric) Shelf 24 to 40 kg
For Use With (Application) Storage of laboratory chemicals
Dimensions (D x W x H) Interior 57 x 60 x 140 cm
Energy Usage 7.6kWh/24 hr.
Frequency 50/60 Hz
Wattage 640 w
Shipping Weight (Metric) 170 kg (gross)