Learn More
SHP Steriltechnik AG Laboklav 100 Wire Mesh Basket
Description
- Wire mesh basket for Laboklav 100
- Stainless Steel
- for horizontal units
- W x H x D (mm): 290 x 145 x 370
Specifications
Specifications
| Capacity (Metric) | 15 L |
| For Use With (Equipment) | Autoclaves, Laboklav 100 |
| Model | Basket, Laboklav 100 |
| Product Type | Wire Mesh Basket |
| Sterility | Non-sterile |