Learn More
Cytiva LabMate™ PD-10 Buffer Reservoirs
Shop All Cytiva Products
missing translation for 'orderingAttributeHoverText'
Quantity:
10 Pack
missing translation for 'unitSize'
Pack of 10
Description
- With Disposable PD-10 Desalting Columns, wash and equilibration buffer can be applied in one step with gravity flow protocol
Specifications
Specifications
| Product Type | Buffer reservoir |
| Quantity | 10 Pack |
| Column Type | Column Accesory |
| For Use With (Application) | High-recovery desalting and Buffer exchange, as well as removal of low-molecular weight compounds packed with Sephadex G-25 medium, makes easier and more convenient to equilibrate PD-10 column |