Learn More
Shamrock™ Labeling Tape Assorted Colors
Click to view available options
Dimensions (LxW):
12.7 x 19 mm
Unit Size:
Pack of 16
Description
- Resistant to water, oil and acid
- Temperature range of -20 to +121°C
- For autoclave, freezer, refrigerator and oven
- Universal marking with ballpoint pen, paper pencil and felt marker
Specifications
Specifications
| Adhesive Type | Removable |
| Color | Assorted |
| Product Type | Label Tape |
| For Use With (Application) | Ideal for color coding, versatile Shamrock labeling tape accepts handwritten notations or can be imprinted to user specifications |
| Quantity | 16 Rolls/Pk. |
| Dimensions (LxW) | 12.7 x 19 mm |
| Length (Metric) | 12.7 m |
| Width (Metric) | 19.05 mm |