Learn More
Hirschmann™ Class AS AR-Glas Volumetric Pipettes
Click to view available options
Volume (Metric):
4 mL
Unit Size:
Pack of 6
Description
- Acc. to DIN EN ISO 648
- One mark, blue graduation
- Conformity certified, with dated batch identification
Specifications
Specifications
| Class | AS |
| Graduated | Yes |
| Graduation Color | Blue |
| Quantity | 6/Pk. |
| Volume (Metric) | 4 mL |
| Type | Labeled Pipet |