Learn More
Shamrock™ Label Slide
Product Code. p-8010111
Click to view available options
Packaging Type:
Roll
Sheet
Unit Size:
Pack of 1000
Pack of 5000
Product Code. 11862631
Brand: Shamrock™ SLSLASER1
Specifications
Type Slide Label
Color White
Dimensions (L x W) 23.8 x 23.8 mm
For Use With (Equipment) Laser Printer
Material Laser
Packaging Type Sheet
Quantity 5000 Pack