Learn More
Shamrock™ Label Slide
Product Code. 11862631
Change view
Click to view available options
Packaging Type:
Roll
Sheet
Unit Size:
Pack of 1000
Pack of 5000
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Packaging Type unitSize
11862631 Sheet Pack of 5000
11832631 Roll Pack of 1000
11872631 - Pack of 5000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11862631 Supplier Shamrock™ Supplier No. SLSLASER1
  • ONLINE EXCLUSIVE PRICE
    112.00€
    60.67€ / Pack of 5000
    Save 51.33€
    45% Off
Estimated Shipment: 22-09-2026
Specifications
Type Slide Label
Color White
Dimensions (L x W) 23.8 x 23.8 mm
For Use With (Equipment) Laser Printer
Material Laser
Packaging Type Sheet
Quantity 5000 Pack