Learn More
Labconco™ Stand-Still™ Isolation Platform
Product Code. p-3831151 Shop All Labconco Corporation Products
Click to view available options
Dimensions (L x W x H):
18.75 x 12 x 1.5 in. (47.6 x 30.5 x 3.81 cm)
Unit Size:
Each
Product Code. 15538854
Brand: Labconco™ 4028200
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
The Labconco Stand-Still™ Isolation Platform is used for supporting microscopes on Purifier™ Cell Logic™+ Biosafety Cabinets with Scope-Ready™ package.
  • Non-porous, autoclavable Type 304 stainless steel base with 1.5 in. (0.4cm) high isolation pads
  • Reduces vertical and horizontal vibrations up to threefold, allowing proper airflow through cabinet
Stand-Still XL Platform:
Requires use of Pure-Vu™ XL Seal 4027810 or 4027811 (Part Numbers)
Specifications
Product Type Vibration Isolation Platform
Color Silver
Autoclavable Autoclavable
Dimensions (L x W x H) 18.75 x 12 x 1.5 in. (47.6 x 30.5 x 3.81 cm)
Dimensions (L x W) 12 x 18.75 in.
For Use With (Application) For supporting microscopes on Cell Logic Biosafety Cabinets with ScopeReady package, to promote proper airflow through the cabinet
For Use With (Equipment) Microscopes, purifier cell logic class II biosafety cabinets
Height (English) 1.5 in.
Height (Metric) 3.81 cm
Length (English) 18.75 in.
Length (Metric) 47.6 cm
Weight (English) 8 lb.
Weight (Metric) 3.6 kg
Width (English) 12 in.
Width (Metric) 30.5 cm
Material Stainless Steel