Learn More
Labconco™ Replacement Lid Gasket
Product Code. 11856775 Shop All Labconco Corporation Products
Change view
Click to view available options
For Use With (Equipment):
Bulk drying chamber
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
11856775 Bulk drying chamber Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11856775 Supplier Labconco™ Supplier No. 7688500
  • ONLINE EXCLUSIVE PRICE
    137.30€
    65.52€ / Pack of 1
    Save 71.78€
    52% Off
 Restricted Item
Estimated Shipment: 17-11-2026
Replacement lids for use with FreeZone 1, 2.5 and 4.5 Liter Freeze Dry Systems.
Replacement lids for use with FreeZone 1, 2.5 & 4.5 Liter Freeze Dry Systems.
Replacement rubber gasket allows the chamber lid of a FreeZone Freeze Dryer to seal to the chamber during the lyophilization process.
Specifications
Type Lid Gasket
Capacity (Metric) 1, 2.5, 4.5 L
For Use With (Equipment) Bulk drying chamber