Learn More
Labconco™ FreeZone™ Freeze-Dry Glass Trap
Product Code. 17806138 Shop All Labconco Corporation Products
Change view
Click to view available options
Material:
Glass
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material Unit Size
17806138 Glass Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17806138 Supplier Labconco™ Supplier No. 7871500
  • ONLINE EXCLUSIVE PRICE
    646.35€
    532.55€ / Pack of 1
    Save 113.80€
    17% Off
 Restricted Item
Estimated Shipment: 20-11-2026
For use in FreeZone freeze dry units
  • Replacement glass trap
  • Fits inside CentriVap Cold Trap collector chamber
  • Protects chamber and lid from corrosive materials
  • Includes insulative cover, 1/2 in. stems and elbows
  • Not compatible with pre-2005 CentriVap Cold Traps
Specifications
Material Glass
For Use With (Equipment) Freezone™ freeze dry units
Type Trap