Learn More
Labconco™ Fast-Freeze™ Flasks, Rubber Tops Only
Shop All Labconco Corporation Products
Click to view available options
Capacity (Metric):
80 mL
300 mL
900 mL
Diameter (Metric) Flask Top Adapter:
1.27 cm
1.9 cm
Unit Size:
Each
Description
Requires
Adapters are required to attach flasks to freeze dry valves–40 and 80mL flasks use a 0.5 in. (1.3cm) adapter; larger sizes use a 0.75 in. (1.9cm)
For use with Fast-Freeze Flasks
- Flexible, noncontaminating silicone rubber top Snaps on and off easily, yet provides a reliable, high-vacuum seal
Specifications
Specifications
| Diameter (Metric) Flask Top Adapter | 1.9 cm |
| Capacity (Metric) | 900 mL |
| Material | Borosilicate Glass |
| Product Type | Flask Top |
| Weight (English) | 1 lb. |
| Disposable | Reusable |
| Mouth Style | Wide |
| Bottom Shape | Flat |
| Diameter (Metric) Outer | 11.1 cm |
| Weight (Metric) | 0.5 kg |