Learn More
Labconco™ Fast-Freeze™ Flask Adapter
Product Code. 15193025 Shop All Labconco Corporation Products
Change view
Click to view available options
Diameter (English):
0.5 in.
0.75 in.
Material:
Borosilicate Glass
Stainless Steel
Style:
45° bend
Straight
Unit Size:
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (English) Material Style unitSize
15193025 0.75 in. Borosilicate Glass Straight Each
11300710 0.5 in. Borosilicate Glass 45° bend Each
10768205 0.5 in. Stainless Steel Straight Each
10763374 0.5 in. Stainless Steel 45° bend Each
10196103 0.75 in. Stainless Steel Straight Each
10604853 0.75 in. Stainless Steel 45° bend Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 15193025 Supplier Labconco™ Supplier No. 7545200
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
Add to basket
Fast-Freeze™ Flask Adapter for connecting Fast-Freeze™ flasks to freeze-dryer valves.
For connecting Fast-Freeze™ flasks to freeze-dryer valves
  • 40 and 80 mL flasks take size 0.5 in. or 0.5 to 0.75 in.
  • Larger flasks fit size 0.75 in. or large end of 0.5 to 0.75 in.
Specifications
Type Flask Adapter
Capacity (Metric) 40, 80 mL
Material Borosilicate Glass
Style Straight
For Use With (Application) For connecting fast freeze™ flasks to freeze-dryer valves
For Use With (Equipment) Fast freeze™ flask
Diameter (English) 0.75 in.
Diameter (Metric) 1.9 cm
Height (English) 0.75 in.
Height (Metric) 1.905 cm
Length (English) 3.25 in.
Length (Metric) 8.26 cm
Weight (English) 0.5 lb.
Weight (Metric) 0.2 kg