Learn More
Labconco™ Fast-Freeze™ Flask Adapter
Product Code. 10763374 Shop All Labconco Corporation Products
Click to view available options
Diameter (English):
0.5 in.
0.75 in.
Material:
Borosilicate Glass
Stainless Steel
Style:
45° bend
Straight
Unit Size:
Each
Product Code. 10763374
Brand: Labconco™ 7547400
Only null left
Add to basket
Add to basket
Fast-Freeze™ Flask Adapter for connecting Fast-Freeze™ flasks to freeze-dryer valves.
For connecting Fast-Freeze™ flasks to freeze-dryer valves
  • 40 and 80 mL flasks take size 0.5 in. or 0.5 to 0.75 in.
  • Larger flasks fit size 0.75 in. or large end of 0.5 to 0.75 in.
Specifications
Type Flask Adapter
Capacity (Metric) 40, 80 mL
Material Stainless Steel
Style 45° bend
For Use With (Application) For connecting fast freeze™ flasks to freeze-dryer valves
For Use With (Equipment) Fast freeze™ flask
Diameter (English) 0.5 in.
Diameter (Metric) 1.27 cm
Height (English) 0.5 in.
Height (Metric) 1.27 cm
Length (English) 5.0 in
Length (Metric) 12.7 cm
Weight (English) 0.5 lb.
Weight (Metric) 0.2 kg