missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ UltraVision™ ONE Detection System: HRP Polymer/DAB Plus Chromogen

Product Code. 12653997 Shop All Epredia Products
Change view
Click to view available options
Quantity:
75 to 150 Tests
Unit Size:
15mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12653997 75 to 150 Tests 15mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12653997 Supplier Epredia™ Supplier No. TL015HDJ

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products

This item is not returnable. View return policy

Employ a one-step polymer system providing increased sensitivity, time savings and detection simplicity with the Epredia™ Lab Vision™ UltraVision™ ONE Detection System: HRP Polymer/DAB Plus Chromogen.

UltraVision ONE is a robust ONE-step polymer system that provides increased sensitivity, time savings and detection simplicity. The UltraVision ONE HRP polymer is an innovative, patented technology. It consists of smaller amino acid polymer subunits that minimize conflicts in binding the target protein. Decreased binding conflicts result in more consistent staining and better signal amplification. Ultimately, this gives the user higher sensitivity and antibody efficiency. UltraVision ONE HRP polymer allows the use of less antibody to obtain better signal-to-noise ratios. This system is also biotin-free, which eliminates background staining found with traditional biotin-based detection methods. For optimal interpretation of results, appropriate positive and negative controls must be included.
  • Specificity: anti-Mouse IgG (H+L), anti-Rabbit IgG (H+L)

  • Enzyme: Peroxidase

  • Chromogen/Substrate: Diaminobenzidine (DAB)

Includes:

TL-015-HDJ: 15mL Ultra V Block (TA-015-UB), 15mL Hydrogen Peroxide Block (TA-015-HP), 15mL UltraVision ONE HRP Polymer (TL-015-PHJ), 15mL DAB Plus Substrate (TA-015-HSX), 1mL DAB Plus Chromogen (TA-001-HCX)

Note:

UltraVision ONE HRP polymer detection system detects rabbit and mouse immunoglobulins bound to an antigen in tissue sections. The specific primary antibody is located by a universal secondary antibody polymer formulation. The amino acid polymer is conjugated to horseradish peroxidase and the Fab fragments of goat anti-rabbit and goat anti-mouse. The polymer complex is then visualized with an appropriate substrate/chromogen.

TRUSTED_SUSTAINABILITY

Specifications

Count 75 to 150 Slides
Quantity 75 to 150 Tests
Includes 15mL Ultra V Block (TA-015-UB), 15mL Hydrogen Peroxide Block (TA-015-HP), 15mL UltraVision ONE HRP Polymer (TL-015-PHJ), 15mL DAB Plus Substrate (TA-015-HSX), 1mL DAB Plus Chromogen (TA-001-HCX)
Chromagen Diaminobenzidine (DAB)
Specificity anti-Mouse IgG (H+L), anti-Rabbit IgG (H+L)
Storage Requirements Store at 2° to 8°C. Each component is stable for 18 months.
Enzyme Peroxidase
Product Line Lab Vision, UltraVision
Substrate Diaminobenzidine (DAB)
compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Germ cell mutagenicity Category 2
Hazard Statement
  • H341-Suspected of causing genetic defects.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: sodium azide, hydrogen peroxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.