Learn More
Lab Vision™ Tris Buffered Saline and Tween 20 (20X), Epredia™
Product Code. 12633977 Shop All Epredia Products
Click to view available options
Quantity:
125 mL
1000 mL
Unit Size:
1 L
125mL
Product Code. 12633977
Brand: Epredia TA125TT
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Take advantage of a wash buffer for immunolabeling techniques with the Utilize Epredia™ Lab Vision™ Tris-Buffered Saline and Tween™ 20 (20X).
  • For in vitro diagnostic use
  • Tween 20 is added to promote more effective washings, resulting in decreased nonspecific background staining
  • Tween 20 aids reagent spreading on slides
Storage Conditions: Store at room temperature. Stable for 2 years from date of manufacture. Use diluted solution within one week
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material TBS with Tween (TBST)
Concentration 20X
For Use With (Application) Anatomical Pathology
Quantity 125 mL
For in vitro diagnostic use