missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epredia™ Lab Vision™ Liquid Fast-Red Substrate System, 1mL Fast Red Chromogen (75X), 60mL Naphthol Phosphate
Shop All Epredia Products
Click to view available options
Substrate Quantity:
60 mL
125 mL
Chromagen Quantity:
1mL
2mL
Unit Size:
125mL
60mL
Description
In the presence of alkaline phosphatase enzyme, Fast-Red produces a bright red reaction product.
- Bright red reaction product can be seen using brightfield or fluorescent microscopy
- Liquid Fast-Red is not soluble in xylenes and alcohols; therefore it may be permanently mounted
Specifications
Specifications
| Chromagen Quantity | 1mL |
| Chromagen Type | Fast Red |
| Product Line | Lab Vision™ |
| Quantity | 1 mL Fast Red Chromogen, 60 mL Naphthol Phosphate |
| Substrate Type | Naphthol Phosphate Substrate |
| Storage Requirements | Store at 2° to 8°C |
| Substrate Quantity | 60 mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction