Learn More
Macherey-Nagel™ LAB-TOP Surface Protection Paper, MN 210 PE
Cover workbenches, table tops and shelves to protect against spills. Macherey-Nagel™ LAB-TOP Surface Protection Paper, MN 210 PE is comprised of quality cellulose filter paper coated on one side with polyethylene to absorb spilled liquids.
Brand:  Macherey-Nagel™ 112010
Product Code. 10129610
  • 361.00€ / Pack of 100
Only null left
In Stock
Description
  • Grade: MN 210 PE
  • Cellulose filter paper (0.22mm thick, 140g/m2), coated on one side with polyethylene
  • Customizable size: roll measures 100m Length x 48cm Width (328 ft. L x 19 in. W)
  • For covering laboratory workbenches, shelves, etc.
  • Especially suited for isotope and bacteriological laboratories and chemical storerooms/cupboards
  • Can also be used to line the cages/pens of small animals and pets
Specifications
Specifications
140 g/m2
Roll
Covering surfaces (benchtop, table, shelf, animal cages) to protect against spills
1 Each
Coated Surface-lining Paper
48 cm
LAB-TOP surface protection paper
Polyethylene-coated Cellulose Paper
MN 210 PE
0.22 mm
100m