Learn More
Carbolite™ Lab Oven
Product Code. 12049129
Change view
Click to view available options
Capacity (Metric):
30 L
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
12049129 30 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12049129 Supplier Carbolite™ Supplier No. LHT4030230SN+TMPR38301
  • ONLINE EXCLUSIVE PRICE
    9320.00€
    7665.00€ / Pack of 1
    Save 1655.00€
    17% Off
 Restricted Item
Estimated Shipment: 04-02-2027
Specifications
Capacity (Metric) 30 L
Type Laboratory Oven
No. of Shelves 2
Length (Metric) 270 mm
Width (Metric) 830 mm
Depth (Metric) 270 mm
Depth (Metric) Interior 305 mm