Learn More
Bochem™ Lab Jack
Product Code. 10505421
Click to view available options
Material:
Anodized Aluminum
Unit Size:
Each
Product Code. 10505421
Brand: Bochem™ 11091
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Connect the blue anodized aluminum Bochem™ Lab Jack to a special adjusting wheel using a ratchet (included).
  • Improved scissor technology with reduced tolerances
  • Ergonomic knob, rounded and user friendly
  • Scissor-construction made of 18/10-stainless steel
Specifications
Material Anodized Aluminum
Length (Metric) Base 200 mm
Width (Metric) Base 200 mm
Includes Ratchet