Learn More
Sperian™ Lab Diagnostics Monovalent Somatic Salmonella Antisera O
Click to view available options
Lens Tint:
Clear
Unit Size:
Each
Specifications
Specifications
| Certifications/Compliance | ISO 9001 / 2000 |
| Lens Coating | Anti-fog |
| Lens Tint | Clear |
| Product Type | Safety goggles |
| For Use With (Application) | Automotive Industry, Building Industry, Chemical Industry |
| Frame Color | Blue |
| Headband Material | Neoprene |
| No. per Pack | 1 Ea. |
| Material | Polycarbonate |
| Frame Material | Rigid Polypropylene Plastic |