Learn More
Novus Biologicals™ L929 CoCl2 treated/Untreated Cell Lysate
Shop All Bio Techne Products
Click to view available options
Quantity:
2 Vials
Unit Size:
Pack of 2
Specifications
Specifications
| Preparation Method | L929 whole cell lysate is provided as a Western blot positive control. The lysates are provided as a set of treated and untreated samples. Use 10 μL / lane for a standard |
| Quantity | 2 Vials |
| Host Species | Mouse |
| Content And Storage | Store at -20°C to -80°C. Avoid freeze/thaw cycles. |
| Target | L929 |