Learn More
L-Valine-d8, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Specifications
Specifications
| Chemical Name or Material | Valine d8 |
| CAS | 35045-72-8 |
| Molecular Formula | C5 2H8 H3 N O2 |
| Quantity | 0.5 g |
| InChI Formula | InChI=1 S/C5H11NO2/c1-3(2)4(6)5(7)8/h3-4 H,6H2,1-2H3,(H,7,8)/t4-/m0/s1/i1D3,2D3,3 D,4 D |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])(C([2 H])([2 H])[2 H])[C@]([2 H])(N)C(=O)O |
| IUPAC Name | (2 S)-2-amino-2,3,4,4,4-pentadeuterio-3-(trideuteriomethyl)butanoic acid |
| Molecular Weight (g/mol) | 125.2 |
| Formula Weight | 125.1292 g/mol |
| Percent Purity | 98 atom % D, min 98% Chemical Purity |