Learn More
L-Valine benzyl ester hydrochloride, 95%
Product Code. p-7041474
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
Product Code. 15498788
Brand: Thermo Scientific Alfa Aesar H62242.06
Request Bulk or Custom Format
CAS: 2462-34-2 | C12H18ClNO2 | 243.73 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2462-34-2
Molecular Formula C12H18ClNO2
Molecular Weight (g/mol) 243.73
MDL Number MFCD22422648
InChI Key ZIUNABFUHGBCMF-MERQFXBCSA-N
Synonym l-valine benzyl ester hydrochloride, val-obzl hcl, h-val-obzl.hcl, s-benzyl 2-amino-3-methylbutanoate hydrochloride, valine benzyl ester hydrochloride, benzyl 2s-2-amino-3-methylbutanoate hydrochloride, val-obzl.hcl, val-obzl. hcl, l-valine benzyl ester hydrochloride
PubChem CID 11601206
IUPAC Name benzyl (2S)-2-amino-3-methylbutanoate;hydrochloride
SMILES Cl.CC(C)[C@H](N)C(=O)OCC1=CC=CC=C1
Specifications
Melting Point 138°C
Quantity 5 g
Formula Weight 243.73
Optical Rotation −10° (c=2 in Water)
Percent Purity 95%
Chemical Name or Material L-Valine benzyl ester hydrochloride