missing translation for 'onlineSavingsMsg'
Learn More

L-Tryptophan (White Crystals or Cryst. Powder), Fisher BioReagents

Product Code. 11887703 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11887703 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11887703

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
This amino acid is suitable for use in tissue culture systems requiring additives.
TRUSTED_SUSTAINABILITY

Specifications

Melting Point 280°C
Ammonium 0.02% max.
CAS 73-22-3
Arsenic (As) 1ppm max.
pH 5.5 to 7.0
Color White
Heavy Metals (as Pb) 10ppm max.
Identification Pass Test
Assay Percent Range 98.5 to 100.5 %
Loss on Drying 0.2% max.
Other Amino Acids Not Detected
Molecular Formula C11H12N2O2
Packaging Glass Bottle
MDL Number MFCD00064340
Residue after Ignition 0.1% max.
Specific Rotation -30 to -33° (+ or -)
Merck Index 15, 9977
State of Solution (%T) 95.0% min.
Synonym l-tryptophan, tryptophan, l-tryptophane, s-tryptophan, tryptophane, h-trp-oh, optimax, trofan, tryptacin, ardeytropin
Alpha Vector Tryptophan
InChI Key QIVBCDIJIAJPQS-VIFPVBQESA-N
SMILES N[C@@H](CC1=CNC2=CC=CC=C12)C(O)=O
IUPAC Name (2S)-2-amino-3-(1H-indol-3-yl)propanoic acid
Molecular Weight (g/mol) 204.23
Quantity 100 g
PubChem CID 6305
ChEBI CHEBI:16828
Formula Weight 204.23
Percent Purity 98.5 to 100.5%
Physical Form Powder
Chemical Name or Material L-Tryptophan
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.