Learn More
L-Threonolactone, TRC
Specifications
Specifications
| Chemical Name or Material | L-Threono-1,4-lactone |
| CAS | 21730-93-8 |
| Molecular Formula | C4 H6 O4 |
| Quantity | 2.5 g |
| Synonym | 2(3H)-Furanone','dihydro-3,4-dihydroxy-','(3R,4S)-, 2(3H)-Furanone','dihydro-3,4-dihydroxy-','(3R-trans)-, L-Threonic acid','γ-lactone (6CI), Threonic acid','γ-lactone','L- (8CI), (3R,4S)-Dihydro-3,4-dihydroxy-2(3H)-furanone, L-Threono-γ-lactone, L-Threonolacton |
| InChI Formula | InChI=1S/C4H6O4/c5-2-1-8-4(7)3(2)6/h2-3,5-6H,1H2/t2-,3+/m0/s1 |
| SMILES | O[C@H]1COC(=O)[C@@H]1O |
| IUPAC Name | (3R,4S)-3,4-dihydroxyoxolan-2-one |
| Molecular Weight (g/mol) | 118.088 |
| Formula Weight | 118.027 |