Learn More
L-Serine Benzyl Ester Hydrochloride, TRC
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Specifications
Specifications
| Chemical Name or Material | L-Serine Benzyl Ester Hydrochloride |
| CAS | 60022-62-0 |
| Molecular Formula | C10H13NO3 . HCl |
| Quantity | 10 mg |
| Synonym | L-Serine','phenylmethyl ester','hydrochloride (1:1) (ACI), L-Serine','phenylmethyl ester','hydrochloride (9CI), Serine benzyl ester hydrochloride (6CI), (S)-Benzyl 2-amino-3-hydroxypropanoate hydrochloride, L-Serine benzyl ester hydrochloride, Benzyl L-serinate hydrochloride, Benzyl (2S)-2-amino-3-hydroxypropanoate hydrochloride |
| InChI Formula | InChI=1S/C10H13NO3.ClH/c11-9(6-12)10(13)14-7-8-4-2-1-3-5-8;/h1-5,9,12H,6-7,11H2;1H/t9-;/m0./s1 |
| SMILES | Cl.N[C@@H](CO)C(=O)OCc1ccccc1 |
| IUPAC Name | benzyl (2S)-2-amino-3-hydroxypropanoate;hydrochloride |
| Molecular Weight (g/mol) | 231.67 |
| Formula Weight | 231.0662 |
Product Content Correction
Product Title
Ser du en mulighed for forbedring?Del en indholdskorrektion