Learn More
Masterflex™ L/S™ Replacement Brushes
Quickly and easily replace components of your Masterflex system
Brand:  Masterflex™ 07520-04
Product Code. 13075656
  • 77.15€ / Pack of 2
Only null left
Estimated Shipment: 27-05-2026
Description
  • Choose replacement brushes and caps based on your Masterflex L/S drive
  • Brushes should be inspected every six months or 2000 hours—whichever comes first
  • Easy to install, these replacement drive parts will help extend the life of your drive
  • Suitable for use with Masterflex L/S series 07520, 07521, 07522, 07523, 07525, 07526, 07527, 07528, 07553 (except -50, -60), 07554 (except -50, -60), 07558, 07567, 07568, 07592, 77300 drives