Learn More
Masterflex™ L/S™ Portable Sampling Pump
Product Code. 16421524 Shop All Masterflex Products
Change view
Click to view available options
Flow Rate:
120 to 1100 mL/min.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Flow Rate Unit Size
16421524 120 to 1100 mL/min. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: This product is under eco-contribution tax.
Product Code. 16421524 Supplier Masterflex™ Supplier No. 0757102
  • ONLINE EXCLUSIVE PRICE
    3025.00€
    2504.51€ / Pack of 1
    Save 520.49€
    17% Off
View all promotions
Estimated Shipment: 07-04-2027
Regulatory: This product is under eco-contribution tax.
Masterflex™ L/S™ Portable Sampling Pump is portable and versatile making it ideal for field applications
  • Features a single-turn potentiometer for speed control, ON indicator, and three-position power/reversing switch
  • Reversible flow and prime/purge function
  • Includes a Masterflex™ L/S™ Easy-Load pump head
  • Also accepts Masterflex™ L/S™ Standard pump heads (order separately)
  • Floats up to 30 minutes if dropped in water when case is closed and latched
  • IP54-rated control panel protects against inclement weather and splashing water
  • Rechargeable battery - ideal for field applications
Specifications
Applications Fluid Transfer, Field Work
Flow Rate 120 to 1100 mL/min.
Pump Tube Length Application Dependent
Enclosure Rating (Nema / IP) IP54-rated Control Panel
Product Type Portable