Learn More
Masterflex™ L/S™ Easy-Load™ II Pump Head Replacement Rotor Assembly
Product Code. 11725169
Click to view available options
For Use With:
Easy-Load II Pump Head 77200-50
Easy-Load II Pump Head 77200-52
Easy-Load II Pump Head 77200-62
Unit Size:
Each
Product Code. 11725169
Brand: Masterflex™ 7720058
Only null left
Add to basket
Add to basket
Masterflex™ L/S™ Easy-Load™ II Pump Head Replacement Rotor Assembly for use with High-Performance Precision Tubing
  • Get the most our of Masterflex™ L/S™ Easy-Load™ II Pump Heads with these replacement parts
  • Choose replacement parts suitable for your L/S™ pump head
  • Replacement of pump head rotor assembly extends the life of pump heads
Specifications
For Use With Easy-Load II Pump Head 77200-50
Material Cold-Rolled Steel
Product Line L/S
Tubing Type L/S™
Type Replacement Pump Head Rotor Assembly