Learn More
Masterflex™ L/S™ Bulk-Packed Platinum-Cured Silicone High-Performance Precision Pump Tubing
Product Code. 11770359
Click to view available options
Tubing Size:
13
14
15
16
17
24
25
35
36
Flow Rate:
1000 mL/min
1100mL/min.
130 mL/min
1700 mL/min
1800 mL/min
2600 mL/min
3400 mL/min
36 mL/min
480 mL/min
Unit Size:
Pack of 10
Pack of 20
Product Code. 11770359
Brand: Masterflex™ 9640436
Currently Unavailable: We have no information when this product will be available.
View alternative products
Excellent biocompatibility that meets multiple classifications
  • Genuine Masterflex™ tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include packs, cases, and spools for customer-friendly ordering and delivery
  • Slightly greater clarity than peroxide-cured tubing
  • Smooth surfaces; lower protein binding levels
  • Fewer potential leachables
  • Pharmaceutical, biotech, peristaltic pumping, and food applications
  • Excellent heat resistance, low compression set, odorless, nontoxic, translucent
  • Comes in convenient cases containing up to 20 sealed bags of tubing (depending on tubing size).
  • Bulk tubing eliminates individual cardboard boxes, reduces unpacking time, and saves you money on shipping.
  • Tubing comes in 25-ft bags.
Specifications
Material Platinum-cured Silicone
Overall Length (English) 25 ft.
Overall Length (Metric) 7.6 m
For Use With (Application) Pharmaceutical, Biotech, Peristaltic Pumping, and Food Applications
Min. Operating Temperature (Metric) -40°C
Tubing Size 36
Flow Rate 3400 mL/min
Hose Barb 3/8 in.
Max. Operating Temperature (Metric) 100°C