Learn More
L-Phenylalanine tert-butyl ester hydrochloride, 98%
Product Code. 15454796
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. 15454796
Brand: Thermo Scientific Alfa Aesar H59122.06
Request Bulk or Custom Format
CAS: 15100-75-1 | C13H20ClNO2 | 257.758 g/mol
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs and food industry. 
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs and food industry. 
Solubility
Soluble in methanol.
Notes
Hygroscopic. Keep away from heat and humidity. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 15100-75-1
Molecular Formula C13H20ClNO2
Molecular Weight (g/mol) 257.758
MDL Number MFCD00038894
InChI Key FDMCEXDXULPJPG-MERQFXBCSA-N
Synonym h-phe-otbu.hcl, h-phe-otbu hcl, tert-butyl 3-phenyl-l-alaninate hydrochloride, s-tert-butyl 2-amino-3-phenylpropanoate hydrochloride, l-phenylalanine tert-butyl ester hydrochloride, l-phenylalanine t-butyl ester hydrochloride, tert-butyl 2s-2-amino-3-phenylpropanoate hydrochloride, phe-otbu.hcl, tert-butyl l-phenylalaninate hydrochloride, phenylalanine, 1,1-dimethylethyl ester, hydrochloride
PubChem CID 11459563
IUPAC Name tert-butyl (2S)-2-amino-3-phenylpropanoate;hydrochloride
SMILES CC(C)(C)OC(=O)C(CC1=CC=CC=C1)N.Cl
Specifications
Melting Point 240°C
Linear Formula C6H5CH2CH(NH2)COOC(CH3)3·HCl
Beilstein 4723424
Solubility Information Soluble in methanol.
Quantity 5 g
Formula Weight 257.76
Optical Rotation +47° (c=2 in Ethanol)
Sensitivity Hygroscopic
Percent Purity 98%
Chemical Name or Material L-Phenylalanine tert-butyl ester hydrochloride