Learn More
L-Phenylalanine-d8, TRC
Specifications
Specifications
| Chemical Name or Material | L-Phenylalanine-d8 |
| CAS | 29909-00-0 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C9 D8 H3 N O2 |
| Quantity | 25 mg |
| Synonym | (2S)-2-Amino-3-phenylpropanoic Acid-d8, (S)-(-)-Phenylalanine-d8, (S)-α-Amino-β-phenylpropionic Acid-d8, (S)-α-Aminohydrocinnamic Acid-d8, (S)-α-Amino-benzenepropanoic Acid-d8, DL-Phenylalanine-D8, DL-Phenyl-d5-alanine-2,3,3-d3 |
| InChI Formula | InChI=1S/C9H11NO2/c10-8(9(11)12)6-7-4-2-1-3-5-7/h1-5,8H,6,10H2,(H,11,12)/i1D,2D,3D,4D,5D,6D2,8D |
| SMILES | [2H]c1c([2H])c([2H])c(c([2H])c1[2H])C([2H])([2H])C([2H])(N)C(=O)O |
| IUPAC Name | 2-amino-2,3,3-trideuterio-3-(2,3,4,5,6-pentadeuteriophenyl)propanoic acid |
| Molecular Weight (g/mol) | 173.24 |
| Show More |