Learn More
L-Norcarnitine, TRC
Product Code. 30425902
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
Unit Size:
100mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30425902 1 g -
30424534 250 mg -
30423541 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30425902 Supplier Toronto Research Chemicals Supplier No. TRCN6614201G
 Restricted Item
Estimated Shipment: 12-11-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2921-13-3
Molecular Formula C6H13NO3
Molecular Weight (g/mol) 147.17
Synonym (R)-4-(Dimethylamino)-3-hydroxybutanoic Acid, L-4-(Dimethylamino)-3-hydroxybutyric Acid, (-)-Norcarnitine
IUPAC Name (R)-4-(dimethylamino)-3-hydroxybutanoic acid
SMILES CN(C)C[C@@H](CC(=O)O)O
Specifications
Quantity 1 g
InChI Formula InChI=1S/C6H13NO3/c1-7(2)4-5(8)3-6(9)10/h5,8H,3-4H2,1-2H3,(H,9,10)/t5-/m1/s1
Formula Weight 147.09
Percent Purity >95%
Chemical Name or Material L-Norcarnitine
Grade HPLC