Learn More
L-Kynurenine, Tocris Bioscience™
Product Code. 18076062
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Product Code. 18076062
Brand: Tocris Bioscience 4393/50
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Tryptophan catabolite; endogenous aryl hydrocarbon receptor ligand
Tryptophan catabolite; endogenous activator of the aryl hydrocarbon receptor (AhR). Immunosuppressant; suppresses allogeneic T cell proliferation.
TRUSTED_SUSTAINABILITY
Specifications
CAS 2922-83-0
Molecular Formula C10H12N2O3
MDL Number MFCD00069912
Quantity 50 mg
Synonym l-kynurenine, s-2-amino-4-2-aminophenyl-4-oxobutanoic acid, kynurenine, l, 3-anthraniloyl-l-alanine, unii-02jw4j5r44, ccris 4425, 2s-2-amino-4-2-aminophenyl-4-oxobutanoic acid, quinurenine, kynurenin, s-alpha,2-diamino-gamma-oxobenzenebutanoic acid
InChI Key YGPSJZOEDVAXAB-QMMMGPOBSA-N
SMILES N[C@@H](CC(=O)C1=CC=CC=C1N)C(O)=O
IUPAC Name (2S)-2-amino-4-(2-aminophenyl)-4-oxobutanoic acid
Molecular Weight (g/mol) 208.22
PubChem CID 161166
ChEBI CHEBI:16946
Formula Weight 208.21
Percent Purity >98%
Chemical Name or Material L-Kynurenine