missing translation for 'onlineSavingsMsg'
Learn More

L(-)-Histidinol dihydrochloride, 99%

Product Code. p-3734174 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 10482611

Brand: Thermo Scientific Acros 228830010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1596-64-1 | C6H13Cl2N3O | 214.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1596-64-1
Molecular Formula C6H13Cl2N3O
Molecular Weight (g/mol) 214.09
MDL Number MFCD00078058
InChI Key FRCAFNBBXRWXQA-XRIGFGBMSA-N
Synonym l-histidinol dihydrochloride, l---histidinol dihydrochloride, beta-aminoimidazole-4-propanol dihydrochloride, l-histidinol 2 hcl, s-2-amino-3-4-imidazolyl propanol dihydrochloride, l--histidinol 2hcl, imidazole-4-propanol, beta-amino-, dihydrochloride, s-beta-amino-1h-imidazole-4-propanol dihydrochloride, s-2-amino-3-1h-imidazol-4-yl propan-1-ol dihydrochloride, 1h-imidazole-4-propanol, beta-amino-, dihydrochloride, s-9ci
PubChem CID 197743
IUPAC Name (2S)-2-amino-3-(1H-imidazol-5-yl)propan-1-ol;dihydrochloride
SMILES C1=C(NC=N1)CC(CO)N.Cl.Cl

Specifications

Infrared Spectrum Authentic
Color White to Beige
Packaging Glass bottle
Specific Rotation − 3.00
Specific Rotation Condition −3° (24°C c=2,H2O)
Melting Point 193.0°C to 199.0°C
Quantity 1 g
Physical Form Crystalline Powder
Assay Percent Range 99%
Formula Weight 214.1
Percent Purity 99%
Chemical Name or Material L(-)-Histidinol dihydrochloride
Show More Show Less
compliance-icons
Product Identifier
  • L(-)-Histidinol dihydrochloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.